missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Antimony tin oxide, NanoTek™, 99.5%

Brand:  Alfa Aesar™ 044930.36

Code : NEW

Additional Details : Weight : 0.50000kg Transport : UN number : 1549 Chem class : 1 Pack group : III

Quantity 500g

 View more versions of this product

Product Code. 11440920

  • £328.70 / 500g


Estimated Shipment date
from Supplier 16-02-2018
Add to basket

Description

Description

  • Moderately conductive particle
  • The incorporation of NanoTek™ antimony tin oxide products in coating systems, films or composites may help in reducing the buildup of static charge
Specifications

Chemical Identifiers

Antimony tin oxide NanoTek™
MFCD00799153
UN1549

Specifications

Antimony tin oxide NanoTek™
99.5%
UN1549
13 to 22nm APS
500g
Powder
MFCD00799153
Sb2O5:SnO2 15:85 wt%
42 to 72m2/g
Videos
SDS