missing translation for 'onlineSavingsMsg'
Learn More

Antimony tin oxide, NanoArc™, 99.5%

Product Code. 11430920
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Quantity unitSize
11430920 100 g 100g
11420920 25 g 25g
11440920 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 11430920 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer 044930.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Moderately conductive particle. The incorporation of NanoTek™ antimony tin oxide products in coating systems, films or composites may help in reducing the buildup of static charge.Antimony tin oxide is used in liquid crystal display, equipment configuration database and light emitting diode. It is used in the glass of train, car and plane, which used to protect against fog and frost. It is used to protect the electromagnetic wave in the radar shielding and protecting area. It is used in the field of optoelectronic display devices, transparent electrode, solar battery and liquid crystal display.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Moderately conductive particle
  • The incorporation of NanoTek™ antimony tin oxide products in coating systems, films or composites may help in reducing the buildup of static charge
TRUSTED_SUSTAINABILITY

Specifikationer

Chemical Name or Material Antimony tin oxide NanoTek™
Physical Form Powder
Quantity 100 g
MDL Number MFCD00799153
UN Number UN1549
Solubility Information Insoluble in water.
Percent Purity 99.5%

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.