missing translation for 'onlineSavingsMsg'
Learn More

Antimony Standard: Sb @ 10000 μg/mL in 1% HNO3, 6% Tartaric Acid, VHG™

Product Code. 17932426
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
Unit Size:
100mL
250mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17932426 100 mL 100mL
17951326 250 mL 250mL
17912256 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17932426 Supplier VHG Labs Supplier No. VHGTSBWTN100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Antimony Standard: Sb @ 10000 μg/mL in 1% HNO3, 6% Tartaric Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Antimony
CAS 7440-36-0
Methods ICP/ICP-MS
Analyte or Component Names Antimony
Shipping Condition Room Temperature
Molecular Formula Sb
Concentration 10000 μg/mL
Certifications/Compliance ISO 17034
Synonym Antimony powder, Stibium metallicum, Antimony, metallic, Antimony, elemental
InChI Formula InChI=1S/Sb
SMILES [Sb]
IUPAC Name antimony
Solvent or Matrix Dilute HNO3
Molecular Weight (g/mol) 121.76
Quantity 100 mL
Formula Weight 120.904 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.