missing translation for 'onlineSavingsMsg'
Learn More

Antimony rod, 12.7mm (0.50in) dia x 10cm (3.9in) long, 99.8% (metals basis), Thermo Scientific Chemicals

Product Code. 15485915
Click to view available options
Quantity:
1 Pc.
5 Pc.
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-36-0
Molecular Formula Sb
Molecular Weight (g/mol) 121.76
MDL Number MFCD00011211 MFCD00134030
InChI Key WATWJIUSRGPENY-UHFFFAOYSA-N
Synonym powder, stibium metallicum, antimony, metallic, antimony, elemental, stibanylidyne, unii-9it35j3uv3, antymonowodor polish, unii-0vkz97k3ub, 0vkz97k3ub, antimonwasserstoffes german
PubChem CID 5354495
ChEBI CHEBI:30304
IUPAC Name antimony
SMILES [Sb]

Specifications

Quantity 1 Pc.
Percent Purity 99.8%
Odor Odorless
Weight ≈81.2g (0.178lbs)/10cm piece
Form Cast Rod
Assay (metals basis)
Chemical Name or Material Antimony rod, 12.7mm (0.50 in.) dia. x 10cm (3.9 in.) long

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'