missing translation for 'onlineSavingsMsg'
Learn More

Antimony, Puriss. p.a., ≥99.995%, Honeywell™

Product Code. 15650520 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15650520 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15650520 Supplier Honeywell Research Chemicals Supplier No. 0063710G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Boiling Point 1635°C
Melting Point 630°C
CAS 7440-36-0
Physical Form Solid
Quantity 10 g
Molecular Formula Sb
MDL Number MFCD00011211 MFCD00134030
UN Number UN2871
Synonym powder, stibium metallicum, antimony, metallic, antimony, elemental, stibanylidyne, unii-9it35j3uv3, antymonowodor polish, unii-0vkz97k3ub, 0vkz97k3ub, antimonwasserstoffes german
InChI Key WATWJIUSRGPENY-UHFFFAOYSA-N
SMILES [Sb]
IUPAC Name antimony
Molecular Weight (g/mol) 121.76
PubChem CID 5354495
ChEBI CHEBI:30304
Formula Weight 121.76g/mol
Percent Purity ≥99.995%
Grade puriss. p.a.
Packaging Glass bottle
Density 6.69g/mL at 25°C
Chemical Name or Material Antimony
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.