missing translation for 'onlineSavingsMsg'
Learn More

Antimony, 99+%, technical, metal, powder, Fisher Chemical

Product Code. 10040020 Shop All Fisher Chemical Products
Click to view available options
:
500g, Plastic powder jar
This item is not returnable. View return policy

Product Code. 10040020

Brand: Fisher Chemical A/7840/53

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Color Gray
CAS 7440-36-0
Melting Point 630°C
Boiling Point 1635°C
Packaging Plastic powder jar
Vapor Pressure 1mmHg 886
Form Solid
Molecular Formula Sb
MDL Number 134030
Formula Weight 121.75g/mol
compliance-icons
Product Identifier
  • Antimony, powder
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.