missing translation for 'onlineSavingsMsg'
Learn More

Antimony powder, -200 mesh, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog Number 11346958
Click to view available options
Quantity:
1 g
5 g
25 g
7440-36-0
Sb
121.76
MFCD00011211 MFCD00134030
WATWJIUSRGPENY-UHFFFAOYSA-N
powder, stibium metallicum, antimony, metallic, antimony, elemental, stibanylidyne, unii-9it35j3uv3, antymonowodor polish, unii-0vkz97k3ub, 0vkz97k3ub, antimonwasserstoffes german
5354495
CHEBI:30304
antimony
[Sb]
7440-36-0
Sb
121.76
MFCD00011211 MFCD00134030
WATWJIUSRGPENY-UHFFFAOYSA-N
powder, stibium metallicum, antimony, metallic, antimony, elemental, stibanylidyne, unii-9it35j3uv3, antymonowodor polish, unii-0vkz97k3ub, 0vkz97k3ub, antimonwasserstoffes german
5354495
CHEBI:30304
antimony
[Sb]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Gray
Appearance Gray powder
Assay Percent Range 99.999% (Metals basis)
Quantity 25 g
Mesh Size -200 Mesh
Odor Odorless
Form Powder
Chemical Name or Material Antimony

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'