missing translation for 'onlineSavingsMsg'
Learn More

Antimony powder, -200 mesh, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog Number 11371138
Click to view available options
Quantity:
10 g
100 g
500 g
2 kg
This item is not returnable. View return policy
7440-36-0
Sb
121.76
MFCD00011211 MFCD00134030
WATWJIUSRGPENY-UHFFFAOYSA-N
powder, stibium metallicum, antimony, metallic, antimony, elemental, stibanylidyne, unii-9it35j3uv3, antymonowodor polish, unii-0vkz97k3ub, 0vkz97k3ub, antimonwasserstoffes german
5354495
CHEBI:30304
[Sb]
This item is not returnable. View return policy
7440-36-0
Sb
121.76
MFCD00011211 MFCD00134030
WATWJIUSRGPENY-UHFFFAOYSA-N
powder, stibium metallicum, antimony, metallic, antimony, elemental, stibanylidyne, unii-9it35j3uv3, antymonowodor polish, unii-0vkz97k3ub, 0vkz97k3ub, antimonwasserstoffes german
5354495
CHEBI:30304
[Sb]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Gray
UN Number UN2871
Quantity 100 g
IUPAC Name distibyne
Percent Purity 99.5%
Mesh Size −200 Mesh
Odor Odorless
Form Powder
Assay (metals basis)
Chemical Name or Material Antimony powder
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
  • H373-May cause damage to organs.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'