missing translation for 'onlineSavingsMsg'
Learn More

Antimony, plasma standard solution, Specpure™, Sb 1000mug/ml, Thermo Scientific Chemicals

£29.90 - £90.92

Chemical Identifiers

Molecular Formula Sb in 30% HCl
MDL Number MFCD00134030
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15427385
View Documents Promotion Details
Thermo Scientific Alfa Aesar
047103.AD
50 mL
£29.90
50mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15437385
View Documents Promotion Details
Thermo Scientific Alfa Aesar
047103.AE
100 mL
£38.90
100mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15447385
View Documents Promotion Details
Thermo Scientific Alfa Aesar
047103.AP
500 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

Sb in 30% HCl
MFCD00134030

Specifications

Sb 1000mug/ml
MFCD00134030
Acidic
Antimony, plasma standard solution, Specpure™
Sb in 30% HCl
UN1789
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H335
Causes severe skin burns and eye damage.
May cause respiratory irritation.

P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c

H290-H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: HYDROCHLORIC ACID

missing translation for 'einecsNumber' : 231-595-7

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only