missing translation for 'onlineSavingsMsg'
Learn More

Antimony, plasma standard solution, Specpure™ Sb 10,000μg/mL, Thermo Scientific Chemicals

Product Code. 11328828
Click to view available options
Quantity:
50 mL
100 mL
500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula Sb in H2O/tartaric acid/tr. HNO3
MDL Number MFCD00134030

Specifications

Concentration Sb 10,000μg/ml
Quantity 50 mL
Physical Form Liquid
Chemical Name or Material Antimony, plasma standard solution, Specpure™

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'