missing translation for 'onlineSavingsMsg'
Learn More

Antimony pentasulfide, pure, Thermo Scientific Chemicals

Product Code. 15274218 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15274218 250 g 250g
15265076 50 g 50g
15264218 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15274218 Supplier Thermo Scientific Acros Supplier No. 330412500
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1315-04-4 | S5Sb2 | 403.82 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1315-04-4
Molecular Formula S5Sb2
Molecular Weight (g/mol) 403.82
MDL Number MFCD00016321
InChI Key PPKVREKQVQREQD-UHFFFAOYSA-N
Synonym antimony pentasulfide, antimony v sulfide, antimony red, antimonic sulfide, antimonial saffron, antimony sulfide sb2s5, antimony sulfide golden, golden antimony sulfide, diantimony pentasulphide, goldschwefel
PubChem CID 16683083
IUPAC Name [bis(sulfanylidene)-$l^{5}-stibanyl]sulfanyl-bis(sulfanylidene)-$l^{5}-stibane
SMILES S=[Sb](=S)S[Sb](=S)=S

Specifications

Quantity 250 g
Linear Formula Sb2S5
Merck Index 15,688
Solubility Information Solubility in water: insoluble.
Formula Weight 403.83
Percent Purity ≥54% (Sb)
Grade Pure
Chemical Name or Material Antimony pentasulfide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.