missing translation for 'onlineSavingsMsg'
Learn More

Antimony, Oil based standard solution, Specpure™ Sb 1000μg/g

Product Code. 15476025
Click to view available options
Quantity:
50 g
Unit Size:
50g
This item is not returnable. View return policy

Product Code. 15476025

Brand: Thermo Scientific Alfa Aesar 014207.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Physical Form Liquid
MDL Number MFCD00134030
Quantity 50 g
Grade Specpure
Chemical Name or Material Antimony Oil based standard solution, Sb 1000μg/g

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.