missing translation for 'onlineSavingsMsg'
Learn More

Antimony(III) sulfide, 99.999% (metals basis)

Product Code. 11391788
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11391788 50 g 50g
11381788 2 g 2g
11371788 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11391788 Supplier Thermo Scientific Alfa Aesar Supplier No. 011683.18
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 1345-04-6 | S3Sb2 | 339.70 g/mol

It is used in glitter compositions, fountain compositions, flash powder and vulcanizing rubber. It is employed in the production of ruby-colored glass and in plastics, as a fire retardant, in explosives, matches, and fireworks. It can be used to prepare a series of thioantimonous salts and Schlippe's salt.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1345-04-6
Molecular Formula S3Sb2
Molecular Weight (g/mol) 339.70
MDL Number MFCD00011217
InChI Key NVWBARWTDVQPJD-UHFFFAOYSA-N
IUPAC Name diantimony(3+) trisulfanediide
SMILES [S--].[S--].[S--].[Sb+3].[Sb+3]

Specifications

Melting Point 550°C
Physical Form Powder
Quantity 50 g
UN Number UN1549
Merck Index 14,713
Solubility Information Insoluble in water,and moderately soluble in acids.
Formula Weight 339.68
Percent Purity 99.999%
Assay (metals basis)
Chemical Name or Material Antimony(III) sulfide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.