missing translation for 'onlineSavingsMsg'
Learn More

Antimony(III) sulfide, 99.9% (metals basis)

Product Code. 11373938
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11373938 50 g 50g
11383938 10 g 10g
11393938 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11373938 Supplier Thermo Scientific Alfa Aesar Supplier No. 040371.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Vacuum deposition | CAS: 1345-04-6 | S3Sb2 | 339.70 g/mol

Antimony Trisulfide is used in the production of pyrotechnics and as a yellow pigment for glass and porcelain. In Vacuum deposition. Used in preparation of Vermilion or yellow pigment, for antimony salts, percussion caps, camouflage paints, and in ruby glass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1345-04-6
Molecular Formula S3Sb2
Molecular Weight (g/mol) 339.70
MDL Number MFCD00011217
InChI Key NVWBARWTDVQPJD-UHFFFAOYSA-N
IUPAC Name diantimony(3+) trisulfanediide
SMILES [S--].[S--].[S--].[Sb+3].[Sb+3]

Specifications

Melting Point 550°C
Physical Form Pieces
Quantity 50 g
UN Number UN1549
Merck Index 14,713
Solubility Information Insoluble in water. Soluble in concentrated acids.
Formula Weight 339.68
Percent Purity 99.9%
Assay (metals basis)
Chemical Name or Material Antimony(III) sulfide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.