missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Antimony(III) sulfide, 98% (metals basis), Thermo Scientific Chemicals
77.33 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
250 g
1 kg
Description
It is manufactured for use in safety matches, military ammunition, explosives and fireworks. It also is used in the production of ruby-colored glass and in plastics as a flame retardant. percussion caps, camouflage paints, ruby glass
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 550°C |
Color | Black |
Physical Form | Powder |
Quantity | 1 kg |
Assay Percent Range | 98.0% min (Metal basis) |
Solubility Information | Insoluble in water. Soluble in HCl with H2S evolution |
Formula Weight | 339.68 |
Odor | Odorless |
Appearance | Black powder |
Chemical Name or Material | Antimony(III) sulfide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'