missing translation for 'onlineSavingsMsg'
Learn More

Antimony(III) oxide, Puratronic™, 99.999% (metals basis)

Product Code. 11340839
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
100 g
Unit Size:
100g
25g
50g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11340839 100 g 100g
11320839 5 g 5g
11330839 25 g 25g
15475955 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11340839 Supplier Thermo Scientific Alfa Aesar Supplier No. 010641.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1309-64-4 | O3Sb2 | 291.52 g/mol

Antimony(III) oxide finds use as flame retardant wherein it synergistically acts with halogenated materials. It is employed in opacifying glasses, ceramics and enamels. In the preparation of optical and ruby glass, antimony trioxide serves as a bubble remover. Further, it is a useful catalyst in the production of polyethylene terephthalate (PET plastic) and the vulcanization of rubber.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1309-64-4
Molecular Formula O3Sb2
Molecular Weight (g/mol) 291.52
MDL Number MFCD00011214
InChI Key GHPGOEFPKIHBNM-UHFFFAOYSA-N
IUPAC Name diantimony(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Sb+3].[Sb+3]

Specifications

Boiling Point 1550°C (sublimation)
Melting Point 656°C
Color White
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.99% (Metals basis)
Solubility Information Very slightly soluble in water; soluble in potassium hydroxide,hydrochloricacid and acetic acid; insoluble in organic solvents.
Formula Weight 291.5
Odor Odorless
Density 5.67
Chemical Name or Material Antimony(III) oxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.