Learn More
Antimony(III) oxide, 99.6% min, Thermo Scientific Chemicals
Chemical Identifiers
CAS | 1309-64-4 |
---|---|
Molecular Formula | O3Sb2 |
Molecular Weight (g/mol) | 291.52 |
MDL Number | MFCD00011214 |
InChI Key | GHPGOEFPKIHBNM-UHFFFAOYSA-N |
Synonym | Antimony trioxide |
IUPAC Name | diantimony(3+) trioxidandiide |
SMILES | [O--].[O--].[O--].[Sb+3].[Sb+3] |
Description
It finds applications in flame retardants, catalysts, batteries, and alloys. It acts as an opacifying agent in glasses, ceramics and enamels. It is used as a catalyst in the manufacture of PET plastic (polyethylene terephthalate), and the vulcanization of rubber.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1309-64-4 | |
291.52 | |
GHPGOEFPKIHBNM-UHFFFAOYSA-N | |
diantimony(3+) trioxidandiide |
O3Sb2 | |
MFCD00011214 | |
Antimony trioxide | |
[O--].[O--].[O--].[Sb+3].[Sb+3] |
Specifications
1550°C (sublimation) | |
1309-64-4 | |
Powder | |
O3Sb2 | |
Antimony trioxide | |
GHPGOEFPKIHBNM-UHFFFAOYSA-N | |
diantimony(3+) trioxidandiide | |
291.5 | |
White powder | |
Antimony(III) oxide |
656°C | |
White | |
99.6% min (Metal basis) | |
MFCD00011214 | |
Poor solubility in water. Slowly dissolves in warm HCl or warm KOH solutions. | |
[O--].[O--].[O--].[Sb+3].[Sb+3] | |
291.52 | |
Odorless | |
5.67 g/cm3 |
Safety and Handling
P201-P202-P281-P308+P313-P501c
H351
missing translation for 'einecsNumber' : 215-175-0
missing translation for 'rtecsNumber' : CC5650000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Antimony(III) oxide
Signal Word
- Warning
Hazard Category
- Carcinogenicity Category 2
Hazard Statement
- H351-Suspected of causing cancer.
Precautionary Statement
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Antimony trioxide, Lead monoxide, Arsenic trioxide
RUO – Research Use Only