missing translation for 'onlineSavingsMsg'
Learn More

Antimony(III) oxide, 99%, Thermo Scientific Chemicals

Catalog Number p-7081233
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy
1309-64-4
O3Sb2
291.52
MFCD00011214
GHPGOEFPKIHBNM-UHFFFAOYSA-N
diantimony(3+) trioxidandiide
[O--].[O--].[O--].[Sb+3].[Sb+3]
This item is not returnable. View return policy
1309-64-4
O3Sb2
291.52
MFCD00011214
GHPGOEFPKIHBNM-UHFFFAOYSA-N
diantimony(3+) trioxidandiide
[O--].[O--].[O--].[Sb+3].[Sb+3]

Antimony(III) oxide is used as a catalyst, vulcanization of rubber, an opacifying agent for glasses, ceramics and enamels. It is used as a flame retardant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Antimony(III) oxide is used as a catalyst, vulcanization of rubber, an opacifying agent for glasses, ceramics and enamels. It is used as a flame retardant.

Solubility
Insoluble in water.

Notes
Store in cool, dry conditions in well sealed containers. Store away from strong oxidizing agents. Keep container tightly closed.
TRUSTED_SUSTAINABILITY
Boiling Point 1550°C (sublimation)
Melting Point 656°C
Quantity 100 g
Merck Index 14,711
Solubility Information Insoluble in water.
Formula Weight 291.5
Percent Purity 99%
Odor Odorless
Density 5.67
Chemical Name or Material Antimony (III) oxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'