missing translation for 'onlineSavingsMsg'
Learn More

Antimony(III) n-butoxide, 99.9% (metals basis), Sb(III) 36.0%, Thermo Scientific™

Product Code. 15446115
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15446115 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15446115 Supplier Thermo Scientific Alfa Aesar Supplier No. 033496.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2155-74-0
Density 1.264 g/mL
Boiling Point 133°C to 135°C (0.4 mmHg)
Flash Point >110°C (230°F)
Molecular Formula C12H27O3Sb
MDL Number MFCD00015238
Quantity 1 g
UN Number UN3082
Sensitivity Moisture sensitive
Synonym antimony iii butoxide, 1-butanol, antimony 3+ salt, antimony iii n-butoxide, tributoxystibine, antimony 3+ tributan-1-olate, acmc-20als1, antimony iii butan-1-olate, antimony 3+ ; butan-1-olate, antimony 3+ ion tris butan-1-olate, antimony 3+ tris butan-1-olate
Solubility Information Soluble in anhydrous butanol, THF
InChI Key YGBFTDQFAKDXBZ-UHFFFAOYSA-N
SMILES CCCC[O-].CCCC[O-].CCCC[O-].[Sb+3]
IUPAC Name antimony(3+);butan-1-olate
Molecular Weight (g/mol) 341.105
PubChem CID 102217
Formula Weight 341.1
Percent Purity 99.9%
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Antimony(III) n-butoxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.