missing translation for 'onlineSavingsMsg'
Learn More

Antimony(III) n-butoxide, 99+%, Thermo Scientific™

Product Code. 11339428
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11339428 10 g 10g
11359428 2 g 2g
11349428 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11339428 Supplier Thermo Scientific Alfa Aesar Supplier No. 043295.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Preparation of coatings, as a catalyst

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2155-74-0
Molecular Formula C12H27O3Sb
Molecular Weight (g/mol) 341.105
MDL Number MFCD00015238
InChI Key YGBFTDQFAKDXBZ-UHFFFAOYSA-N
Synonym antimony iii butoxide, 1-butanol, antimony 3+ salt, antimony iii n-butoxide, tributoxystibine, antimony 3+ tributan-1-olate, acmc-20als1, antimony iii butan-1-olate, antimony 3+ ; butan-1-olate, antimony 3+ ion tris butan-1-olate, antimony 3+ tris butan-1-olate
PubChem CID 102217
IUPAC Name antimony(3+);butan-1-olate
SMILES CCCC[O-].CCCC[O-].CCCC[O-].[Sb+3]

Specifications

Density 1.264 g/mL
Boiling Point 133°C to 135°C (0.4 mmHg)
Flash Point >110°C (230°F)
Quantity 10 g
UN Number UN3082
Sensitivity Moisture sensitive
Solubility Information Soluble in anhydrous butanol
Formula Weight 341.1
Percent Purity ≥99%
Physical Form Liquid
Chemical Name or Material Antimony (III) n-butoxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.