missing translation for 'onlineSavingsMsg'
Learn More

Antimony(III) isopropoxide, 99.9% (metals basis)

Product Code. 11348028
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11348028 25 g 25g
11358028 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11348028 Supplier Thermo Scientific Alfa Aesar Supplier No. 036589.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 18770-47-3 | C9H21O3Sb | 299.024 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18770-47-3
Molecular Formula C9H21O3Sb
Molecular Weight (g/mol) 299.024
MDL Number MFCD00143623
InChI Key HYPTXUAFIRUIRD-UHFFFAOYSA-N
Synonym antimony iii isopropoxide, antimony 3+ tripropan-2-olate, triisopropoxyantimony, acmc-209epx, antimony iii propan-2-olate, antimony 3+ ; propan-2-olate, 2-propanol,antimony 3+ salt 9ci, antimony 3+ ion tris propan-2-olate, antimony 3+ tris propan-2-olate
PubChem CID 3034303
IUPAC Name antimony(3+);propan-2-olate
SMILES CC(C)[O-].CC(C)[O-].CC(C)[O-].[Sb+3]

Specifications

Color Colorless
Density 1.31 g/mL
Boiling Point 68°C (3 mmHg)
Flash Point 77°C (141°F)
Quantity 25 g
UN Number UN3082
Sensitivity Moisture sensitive
Formula Weight 298.8
Percent Purity 99.9%
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Antimony(III) isopropoxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.