missing translation for 'onlineSavingsMsg'
Learn More

Antimony(III) iodide, ultra dry, 99.998% (metals basis)

Product Code. 11363837
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11363837 5 g 5g
11495040 1 g 1g
11373837 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11363837 Supplier Thermo Scientific Alfa Aesar Supplier No. 035676.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 7790-44-5 | I3Sb | 502.47 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-44-5
Molecular Formula I3Sb
Molecular Weight (g/mol) 502.47
MDL Number MFCD00016322
InChI Key KWQLUUQBTAXYCB-UHFFFAOYSA-K
Synonym antimony iii iodide, stibine, triiodo, antimony triiodide, antimon iii iodid, antimony iodide sbi3, antimony iodide, antimony iii iodide, ultra dry, antimony iii triiodide, hsdb 437, triiodostibine #
PubChem CID 24630
SMILES [Sb+3].[I-].[I-].[I-]

Specifications

Boiling Point 420°C
Melting Point 168°C
Physical Form Beads
Quantity 5 g
UN Number UN3260
Merck Index 14,710
Solubility Information Decomposed by water to SbOI
IUPAC Name antimony(3+) triiodide
Formula Weight 502.46
Percent Purity 99.998%
Odor Acrid
Packaging Ampule
Sensitivity Moisture Sensitive
Assay (metals basis)
Chemical Name or Material Antimony(III) iodide, Ultra dry
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.