missing translation for 'onlineSavingsMsg'
Learn More

Antimony(III) fluoride, 98%

Product Code. 11427996
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11427996 50 g 50g
11437996 250 g 250g
11417996 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11427996 Supplier Thermo Scientific Alfa Aesar Supplier No. A14068.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7783-56-4 | F3Sb | 178.755 g/mol

Antimony(III) fluoride is used as a reagent in inorganic and organofluorine chemistry. It acts as a fluorination reagent and electroplating agent. Further, it is used in the preparation of dye, pottery, porcelains, ceramic enamels and glazes. It is utilized in the preparation of freon.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Antimony(III) fluoride is used as a reagent in inorganic and organofluorine chemistry. It acts as a fluorination reagent and electroplating agent. Further, it is used in the preparation of dye, pottery, porcelains, ceramic enamels and glazes. It is utilized in the preparation of freon.

Solubility
Soluble in water, methanol and acetone. Insoluble in ammonia.

Notes
Moisture sensitive. It is incompatible with acids, perchloric acid, water and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-56-4
Molecular Formula F3Sb
Molecular Weight (g/mol) 178.76
MDL Number MFCD00011218
InChI Key GUNJVIDCYZYFGV-UHFFFAOYSA-K
Synonym antimony trifluoride, trifluorostibine, antimony fluoride, antimony iii fluoride, stibine, trifluoro, trifluoroantimony, antimoine fluorure, antimonous fluoride, antimoine fluorure french, hsdb 438
PubChem CID 24554
IUPAC Name trifluorostibane
SMILES F[Sb](F)F

Specifications

Boiling Point 376°C
Melting Point 292°C
Quantity 50 g
UN Number UN2923
Merck Index 14,709
Solubility Information Soluble in water,methanol and acetone. Insoluble in ammonia.
Formula Weight 178.75
Percent Purity 98%
Odor Odorless
Sensitivity Moisture sensitive
Density 4.379
Chemical Name or Material Antimony(III) fluoride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.