missing translation for 'onlineSavingsMsg'
Learn More

Antimony(III) ethoxide, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog Number 11374758
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
10433-06-4
C6H15O3Sb
256.943
MFCD00015123
JGOJQVLHSPGMOC-UHFFFAOYSA-N
antimony triethoxide, triethoxyantimony, acmc-20alrx, sb oet 3, antimony iii ethoxide, antimony 3+ ; ethanolate, ethanol, antimony 3+ salt 9ci, antimony 3+ tris ethoxide, antimony 3+ ion tris ethoxide
66326
antimony(3+);ethanolate
CC[O-].CC[O-].CC[O-].[Sb+3]
This item is not returnable. View return policy
10433-06-4
C6H15O3Sb
256.943
MFCD00015123
JGOJQVLHSPGMOC-UHFFFAOYSA-N
antimony triethoxide, triethoxyantimony, acmc-20alrx, sb oet 3, antimony iii ethoxide, antimony 3+ ; ethanolate, ethanol, antimony 3+ salt 9ci, antimony 3+ tris ethoxide, antimony 3+ ion tris ethoxide
66326
antimony(3+);ethanolate
CC[O-].CC[O-].CC[O-].[Sb+3]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.524 g/mL
Boiling Point 95°C (11 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
UN Number UN3082
Sensitivity Moisture sensitive
Formula Weight 256.9
Percent Purity 99.9%
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Antimony (III) ethoxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'