missing translation for 'onlineSavingsMsg'
Learn More

Antimony(Iii) Chloride, puriss. p.a.ACS Reagent, ≥99.0% (RT), Honeywell Fluka™

Product Code. 15660980 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15660980 100 g Poly Bottle 100g
15670980 500 g Poly Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15660980 Supplier Honeywell-Fluka Supplier No. 10774100G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Puriss. p.a., ACS reagent,≥99.0% (RT)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-91-9
Molecular Formula Cl3Sb
Molecular Weight (g/mol) 228.11
MDL Number MFCD00011212
InChI Key FAPDDOBMIUGHIN-UHFFFAOYSA-K
Synonym antimony trichloride, trichlorostibine, antimony chloride, antimony iii chloride, stibine, trichloro, antimontrichlorid, antimonous chloride, butter of antimony, antimony butter, caustic antimony
PubChem CID 24814
ChEBI CHEBI:74856
IUPAC Name trichlorostibane
SMILES [Cl-].[Cl-].[Cl-].[Sb+3]

Specifications

Boiling Point 283°C
Melting Point 73.4°C
Color White
Quantity 100 g
Linear Formula SbCl3
UN Number UN1733
Formula Weight 228.12g/mol
Percent Purity ≥99.0% (RT)
Grade ACS/puriss. p.a.
Packaging Poly Bottle
Appearance Crystals/Powder
Vapor Pressure 1mmHg (49°C)
Density 3.140g/cu.cm; Vapor Density: 7.9 (vs air)
Chemical Name or Material Antimony(Iii) Chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.