missing translation for 'onlineSavingsMsg'
Learn More

Antimony(III) chloride, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. p-7051060
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
This item is not returnable. View return policy

Product Code. 11303948

Brand: Thermo Scientific Alfa Aesar 040372.04

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Antimony(III) chloride is used as a chlorinating agent, as a fireproofing agent in textiles; in bronzing steel and as a mordant in dyeing as well as a caustic in medicine. It is also used as an apoptosis agent that is used to detect cholesterols and carotenoids. It is also employed to detect certain compounds, such as cholesterols and carotenoids. This agent seems to cause dehydration of cholesterol to 3,5-cholestadiene and 2,4-cholestadiene which produce the blue color seen during the reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-91-9
Molecular Formula Cl3Sb
Molecular Weight (g/mol) 228.11
MDL Number MFCD00011212
InChI Key FAPDDOBMIUGHIN-UHFFFAOYSA-K
Synonym antimony trichloride, trichlorostibine, antimony chloride, antimony iii chloride, stibine, trichloro, antimontrichlorid, antimonous chloride, butter of antimony, antimony butter, caustic antimony
PubChem CID 24814
ChEBI CHEBI:74856
SMILES [Cl-].[Cl-].[Cl-].[Sb+3]

Specifications

Boiling Point 223.5°C
Melting Point 73.4°C
Physical Form Crystalline Aggregates
Quantity 2 g
UN Number UN1733
Merck Index 14,707
Solubility Information Soluble in alcohol and acetone and acids.
IUPAC Name trichlorostibane
Formula Weight 228.11
Percent Purity 99.999%
Grade Puratronic
Odor Pungent
Sensitivity Moisture sensitive
Density 3.14 g/mL
Assay (metals basis)
Chemical Name or Material Antimony(III) chloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.