missing translation for 'onlineSavingsMsg'
Learn More

Antimony(III) chloride, ACS, 99.0% min

Product Code. 11383348
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11383348 1 kg 1kg
11373348 50 g 50g
11393348 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11383348 Supplier Thermo Scientific Alfa Aesar Supplier No. 036281.A1
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10025-91-9 | Cl3Sb | 228.11 g/mol

Antimony(III) chloride is engaged to detect certain compounds, such as cholesterols and carotenoids. It is likewise employed as a dye mordant and a fabric flame retardant. It is used as inorganic and organic chlorination catalyst. It is used to coat the steel to make bronze, used as a cutting material of antimony salt and as a preservative in medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-91-9
Molecular Formula Cl3Sb
Molecular Weight (g/mol) 228.11
MDL Number MFCD00011212
InChI Key FAPDDOBMIUGHIN-UHFFFAOYSA-K
Synonym antimony trichloride, trichlorostibine, antimony chloride, antimony iii chloride, stibine, trichloro, antimontrichlorid, antimonous chloride, butter of antimony, antimony butter, caustic antimony
PubChem CID 24814
ChEBI CHEBI:74856
IUPAC Name trichlorostibane
SMILES [Cl-].[Cl-].[Cl-].[Sb+3]

Specifications

Boiling Point 223.5°C
Melting Point 73.4°C
Color White
Physical Form Crystalline
Quantity 1 kg
Assay Percent Range 99% min.
Solubility Information Soluble in water,HCl solution,alcohol,benzene,carbon disulfide,dioxane,chloroform,ether,acetone,carbon tetrachloride. Insoluble in organic bases.
Formula Weight 228.11
Grade ACS
Odor Pungent
Sensitivity Moisture sensitive
Appearance White crystalline
Density 3.14
Chemical Name or Material Antimony(III) chloride, For analysis ACS
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.