missing translation for 'onlineSavingsMsg'
Learn More

Antimony(III) bromide, 99.5% (metals basis)

Product Code. 11388688
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11388688 10 g 10g
11398688 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11388688 Supplier Thermo Scientific Alfa Aesar Supplier No. 041574.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7789-61-9 | Br3Sb | 361.472 g/mol

Antimony(III) bromide is used in the production of other antimony compounds, in chemical analysis, as a mordant, and in dyeing. It can be added to polymers such as polyethylene as a fire retardant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-61-9
Molecular Formula Br3Sb
Molecular Weight (g/mol) 361.472
MDL Number MFCD00016317
InChI Key RPJGYLSSECYURW-UHFFFAOYSA-K
Synonym antimony tribromide, antimony iii bromide, antimony bromide, tribromostibine, stibine, tribromo, antimonous bromide, antimony bromide sbbr3, unii-6pm239qd86, hsdb 441, sbbr3
PubChem CID 24615
IUPAC Name tribromostibane
SMILES Br[Sb](Br)Br

Specifications

Boiling Point 280°C
Melting Point 96.6°C
Quantity 10 g
UN Number UN3260
Merck Index 14,706
Solubility Information Soluble in dilute hydrochloric acid,hydrobromic acid,carbon disulfide,alcohol,acetone,ammonia,benzene,chloroform.
Formula Weight 361.48
Percent Purity 99.5%
Refractive Index 1.74
Sensitivity Moisture sensitive
Density 4.148 g/mL
Assay (metals basis)
Chemical Name or Material Antimony(III) bromide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Antimony (III) bromide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Antimony tribromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.