missing translation for 'onlineSavingsMsg'
Learn More

Antimony(III) acetate, 97%

Product Code. 11355009
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
Unit Size:
100g
10g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11355009 500 g 500g
11345009 10 g 10g
11365009 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11355009 Supplier Thermo Scientific Alfa Aesar Supplier No. 087657.36
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6923-52-0 | C6H9O6Sb | 298.892 g/mol

Antimony(III) acetate is a catalyst used in the production of synthetic fibers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6923-52-0
Molecular Formula C6H9O6Sb
Molecular Weight (g/mol) 298.892
MDL Number MFCD00014974
InChI Key JVLRYPRBKSMEBF-UHFFFAOYSA-K
Synonym antimony triacetate, antimony iii acetate, antimony acetate, acetic acid, antimony 3+ salt, octan antimonity czech, antimony 3+ triacetate, acetic acid, antimony 3+ salt 3:1, acetic acid, trianhydride with antimonic acid h3sbo3, acetic acid, antimony salt, octan antimonity
PubChem CID 23354
IUPAC Name antimony(3+);triacetate
SMILES CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].[Sb+3]

Specifications

Quantity 500 g
UN Number UN1549
Sensitivity Moisture sensitive
Solubility Information Decomposes in water
Formula Weight 298.88
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Antimony (III) acetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.