missing translation for 'onlineSavingsMsg'
Learn More

Antimony for AAS 0.02mg/L matrix HNO3 diluted, Fisher Chemical™
BIOPHARMA

Product Code. 12924545 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12924545 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12924545 Supplier Fisher Chemical Supplier No. E 050 050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Bottle
CAS 7732-18-5
Physical Form Liquid
Molecular Formula Sb
Concentration 0.02 mg/L
Type Flame Single-element Standard Solution
Solvent or Matrix 1% HNO3 (diluted)
Quantity 50 mL
Chemical Name or Material Antimony
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.