missing translation for 'onlineSavingsMsg'
Learn More

Antide acetate, Thermo Scientific Chemicals

Product Code. p-7043265
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
This item is not returnable. View return policy

Product Code. 15418149

Brand: Thermo Scientific Alfa Aesar J66692.M^

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112568-12-4
Molecular Formula C82H108ClN17O14
Molecular Weight (g/mol) 1591.32
MDL Number MFCD00133104,MFCD00133104
InChI Key QRYFGTULTGLGHU-NBERXCRTSA-N
Synonym antide, iturelix, antide acetate, nal-lys-gnrh, unii-94055uoq3y, nal-lys-gnrhant, d-alaninamide, n-acetyl-3-2-naphthalenyl-d-alanyl-4-chloro-d-phenylalanyl-3-3-pyridinyl-d-alanyl-l-seryl-n6-3-pyridinylcarbonyl-l-lysyl-n6-3-pyridinylcarbonyl-d-lysyl-l-leucyl-n6-1-methylethyl-l-lysyl-l-prolyl, n-acetyl-3-naphhalen-2-yl-d-alanyl-p-chloro-d-phenylalanyl-3-3-pyridyl-d-alanyl-l-seryl-n sup 6-nicotinyl-l-lysyl-n sup 6-nicotinyl-d-lysyl-l-leucyl-n sup 6-isopropyl-l-lysyl-l-prolyl-d-alaninamide, d-alaninamide, n-acetyl-3-2-naphthalenyl-d-alanyl-4-chloro-d-phenylalanyl-3-3-pyridinyl-d-alanyl-l-seryl-n sup 6-3-pyridinylcarbonyl-l-lysyl-n sup 6-3-pyridinylcarbonyl-d-lysyl-l-leucyl-n sup 6-1-methylethyl-l-lysyl-l-prolyl, iturelix usan:inn
PubChem CID 16130938
IUPAC Name (2S)-N-[(1R)-1-{[(1S)-1-{[(2S)-1-[(2S)-2-{[(1R)-1-carbamoylethyl]carbamoyl}pyrrolidin-1-yl]-1-oxo-6-[(propan-2-yl)amino]hexan-2-yl]carbamoyl}-3-methylbutyl]carbamoyl}-5-[(pyridin-3-yl)formamido]pentyl]-2-[(2S)-2-[(2R)-2-[(2R)-3-(4-chlorophenyl)-2-[(2R)-2-acetamido-3-(naphthalen-2-yl)propanamido]propanamido]-3-(pyridin-3-yl)propanamido]-3-hydroxypropanamido]-6-[(pyridin-3-yl)formamido]hexanamide
SMILES CC(C)C[C@H](NC(=O)[C@@H](CCCCNC(=O)C1=CC=CN=C1)NC(=O)[C@H](CCCCNC(=O)C1=CC=CN=C1)NC(=O)[C@H](CO)NC(=O)[C@@H](CC1=CC=CN=C1)NC(=O)[C@@H](CC1=CC=C(Cl)C=C1)NC(=O)[C@@H](CC1=CC=C2C=CC=CC2=C1)NC(C)=O)C(=O)N[C@@H](CCCCNC(C)C)C(=O)N1CCC[C@H]1C(=O)N[C@H](C)C(N)=O

Specifications

Color White
Quantity 1 mg
Formula Weight 1591.3
Physical Form Solid
Chemical Name or Material Antide acetate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.