missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNHIT3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA543577
Les retours ne sont pas autorisés pour ce produit.
Consulta la politica di reso
Descrizione
Peptide sequence: HKEQCNPETR PVEKKIRSAL PTKTVKPVEN KDDDDSIADF LNSDEEEDRV Sequence homology: Dog: 93%; Horse: 93%; Human: 100%; Pig: 100%; Rabbit: 93%.
ZNHIT3 contains 1 HIT-type zinc finger. The exact function of ZNHIT3 remains unknown.
Specifica
| ZNHIT3 | |
| Polyclonal | |
| Unconjugated | |
| ZNHIT3 | |
| HNF-4a coactivator; Thyroid hormone receptor interactor 3; thyroid receptor interacting protein 3; thyroid receptor-interacting protein 3; TR-interacting protein 3; TRIP3; TRIP-3; Zinc finger HIT domain-containing protein 3; zinc finger HIT domain-containing protein-like protein; zinc finger HIT type 3; zinc finger HIT-type containing 3; zinc finger, HIT domain containing 3; zinc finger, HIT type 3; zinc finger, HIT-type containing 3; ZNHIT3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 9326 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q15649 | |
| ZNHIT3 | |
| Synthetic peptide directed towards the N-terminal of human ZNHIT3 (aa 38-87). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto