missing translation for 'onlineSavingsMsg'
Learn More

zinc finger protein 83 (A01), Mouse anti-Human, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a partial recombinant ZNF83.

Brand:  Abnova H00055769-A01.50uL

Product Code. 16168356

  • £209.00 / 50µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Sequence: MHGRKDDAQKQPVKNQLGLNPQSHLPELQLFQAEGKIYKYDHMEKSVNSSSLVSPPQRISSTVKTHISHTYECNFVDSLFTQKEKANIGTEHYKCNERGKAFHQGLHFTIH
Specifications

Specifications

zinc finger protein 83
Polyclonal
Mouse polyclonal antibody raised against a partial recombinant ZNF83.
ZNF83
FLJ11015/FLJ14876/FLJ30097/FLJ90585/HPF1/MGC33853/ZNF816B
Mouse
50 μL
Transcription Regulation
Primary
Human, Mouse
Serum
ELISA, Western Blot
Unconjugated
50% glycerol
NM_018300
ZNF83
ZNF83 (NP_060770, 1 a.a. ∼ 111 a.a) partial recombinant protein with GST tag.
RUO
Yes
55769
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.