missing translation for 'onlineSavingsMsg'
Learn More
Learn More
zinc finger protein 563, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human ZNF563 protein.
Brand: Abnova H00147837-B01P
This item is not returnable.
View return policy
Description
Sequence: MDAVAFEDVAVNFTQEEWALLGPSQKNLYRYVMQETIRNLDCIRMIWEEQNTEDQYKNPRRNLRCHMVERFSESKDSSQCGETFSLIRDSIVNNSICPGEDPCQSAECEEVIMGHLSLNSHIRVDSGHKPHEYQEYGEKPHTHKQRGKAFSYHHSFQSRGRPHTGKKRYECKECGKTFSSRRNLRRHMVVQGGNRPYKCKLCGKAFFWPSLLRMHERTHTGEKPYECKQCSKAFPFYSSYRRHERMHTGEKPYECKQCSKALPDSSSYIRHERTHTGEKPYTCKQCGKAFSVSSSLRRHETTHSAEKPYECKQCGKTFHHLGSFQIHMKRHTGDRPHKCKICGKGFDRPSLVRYHERIHTGEKPYECKQCGKTLSHSSSFRRHMIMHTGGGPHKCKICGKAFVYPSVCQRHEKSHSGEKPYECKQCGKALSHSSSFRRHMVMHTGDGPNKCKVCGKAFVYPSVCQRHEKTHWRETISpecifications
| zinc finger protein 563 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human ZNF563 protein. | |
| ZNF563 | |
| FLJ34797 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| 147837 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| Immunofluorescence, Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_145276.1 | |
| ZNF563 | |
| ZNF563 (NP_660319.1, 1 a.a. ∼ 476 a.a) full-length human protein. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction