missing translation for 'onlineSavingsMsg'
Learn More

zinc finger protein 274, Mouse, Clone: 2F5, Abnova™

Product Code. 16183176
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16183176 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16183176 Supplier Abnova Supplier No. H00010782M02.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse monoclonal antibody raised against a partial recombinant ZNF274.

This gene encodes a zinc finger protein containing five C2H2-type zinc finger domains, one or two Kruppel-associated box A (KRAB A) domains, and a leucine-rich domain. The encoded protein has been suggested to be a transcriptional repressor. It localizes predominantly to the nucleolus. Alternatively spliced transcript variants encoding different isoforms exist. These variants utilize alternative polyadenylation signals. [provided by RefSeq

Sequence: QKIDNPESQANSGALDTNQVLLHKIPPRKRLRKRDSQVKSMKHNSRVKIHQKSCERQKAKEGNGCRKTFSRSTKQITFIRIHKGSQVCRCSECGKIFRNPRYFSVHKKIHT

Specifications

Antigen zinc finger protein 274
Applications ELISA, Immunofluorescence
Classification Monoclonal
Clone 2F5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant ZNF274.
Formulation PBS with no preservative; pH 7.4
Gene ZNF274
Gene Accession No. NM_133502
Gene Alias DKFZp686K08243/FLJ37843/HFB101/ZF2/ZKSCAN19
Gene Symbols ZNF274
Host Species Mouse
Immunogen ZNF274 (NP_598009, 420 a.a. ∼ 530 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Transcription Regulation
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 10782
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.