missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF157 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA527966
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431, H1299, HeLaS3, Raji. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene product is a likely zinc finger family transcription factor. It contains KRAB-A and KRAB-B domains that act as transcriptional repressors in related proteins, and multiple zinc finger DNA binding motifs and finger linking regions characteristic of the Kruppel family. This gene is part of a gene cluster on chromosome Xp11.23.
Specifications
| ZNF157 | |
| Polyclonal | |
| Unconjugated | |
| ZNF157 | |
| HZF22; Zinc finger protein 157; zinc finger protein 22; zinc finger protein HZF22; ZNF157 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7712 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.45 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| P51786 | |
| ZNF157 | |
| Recombinant protein encompassing a sequence within the center region of human ZNF157. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction