missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF143 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA521448
This item is not returnable.
View return policy
Description
Recommended positive controls: HepG2. Predicted reactivity: Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Transcriptional activator. Activates the gene for selenocysteine tRNA (tRNAsec). Binds to the SPH motif of small nuclear RNA (snRNA) gene promoters. Participates in efficient U6 RNA polymerase III transcription via its interaction with CHD8.
Specifications
| ZNF143 | |
| Polyclonal | |
| Unconjugated | |
| ZNF143 | |
| AA959806; AU015869; D7Ertd805e; hStaf; KRAB14; mStaf; pHZ-1; SBF; selenocysteine tRNA activating factor; selenocysteine tRNA gene transcription activating factor; selenocysteine tRNA gene transcription-activating factor; si:dkey-8l13.2; SPH-binding factor; staf; transcriptional activator Staf; Zfp143; zfp-143; Zfp79; Zfp80-rs1; zgc:66276; zinc finger protein 143; zinc finger protein 143b; zinc finger protein 80, related sequence 1; znf143; znf143b | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7702 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 0.83 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| P52747 | |
| ZNF143 | |
| Recombinant protein encompassing a sequence within the C-terminus region of human ZNF143. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction