missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZFP36 Polyclonal Antibody
ZFP36 Polyclonal Antibody
Brand: Invitrogen PA541450
This item is not returnable.
View return policy
Description
ZFP36 Polyclonal Antibody for Western Blot, IHC (P)
Specifications
| ZFP36 | |
| Polyclonal | |
| Unconjugated | |
| Zfp36 | |
| G0/G1 switch regulatory protein 24; G0S24; GOS24; growth factor-inducible nuclear protein NUP475; mRNA decay activator protein ZFP36; Nup475; RNF162A; Tax interacting protein; TIS11; TIS11A; TIS11D; TISII; TNF alpha inhibitor; TPA-induced sequence 11; Tristetraprolin; tristetraprolin; LOW QUALITY PROTEIN: mRNA decay activator protein ZFP36; tristetraproline; TTP; Zfp36; zfp-36; ZFP36 ring finger protein; zinc finger protein 36; zinc finger protein 36 homolog; zinc finger protein 36, C3H type, homolog; zinc finger protein, C3H type, 36 homolog | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 7538 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P26651 | |
| Zfp36 | |
| Synthetic peptide directed towards the middle region of human ZFP36. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction