missing translation for 'onlineSavingsMsg'
Learn More

Zfp110 Polyclonal Antibody, Invitrogen™

Product Code. 11581412
Change view
Click to view available options
Quantity:
500 μg
Unit Size:
500µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11581412 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11581412 Supplier Invitrogen Supplier No. OSZ00002G

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Reconstitute with 500 μL of distilled water.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Zfp110
Applications Immunohistochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation Antibody with no preservative
Gene Zfp110
Gene Accession No. Q923B3, Q4V8E9
Gene Alias Zfp110
Gene Symbols Zfp110
Host Species Rabbit
Immunogen A synthetic peptide from c-terminal region of rat zinc finger protein 110
Purification Method Ammonium sulfate precipitation
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 308362, 65020
Target Species Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.