Learn More
ZFHX1B, Mouse, Clone: 2D12, Abnova™
Mouse monoclonal antibody raised against a partial recombinant ZFHX1B.
Brand: Abnova H00009839-M03.100ug
Description
The SMADIP1 gene (also known as SIP1) is a member of the delta-EF1 (ZEB1; MIM 189909)/Zfh1 family of 2-handed zinc finger/homeodomain proteins. SMADIP1 interacts with receptor-mediated, activated full-length SMADs (see MIM 605568) (Verschueren et al., 1999 [PubMed 10400677]).[supplied by OMIM
Sequence: SITPQGYSDSEERESMPRDGESEKEHEKEGEDGYGKLGRQDGDEEFEEEEEESENKSMDTDPETIRDEEETGDHSMDDSSEDGKMETKSDHEEDNMEDGMSpecifications
| ZFHX1B, | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_014795.2 | |
| ZEB2 | |
| ZFHX1B (NP_055610.1, 1115 a.a. ∼ 1214 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Immunofluorescence, Western Blot | |
| 2D12 | |
| Mouse monoclonal antibody raised against a partial recombinant ZFHX1B. | |
| ZEB2 | |
| KIAA0569/SIP-1/SIP1/SMADIP1/ZFHX1B | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 9839 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.