Learn More
Zap70 (pY292) Mouse anti-Human, Alexa Fluor 488, Clone: J34-602, BD
Mouse Monoclonal Antibody
Brand:  BD Biosciences 558516
Product Code. 15869448
  • ONLINE EXCLUSIVE PRICE
    £355.00
    £340.86 / Each
    Save £14.14
    3% Off
Description
The 70kDa ζchain-associated protein (ZAP70) is a Syk-family protein tyrosine kinase (PTK) that associates with the ζ subunit of the T cell antigen receptor (TCR) and undergoes tyrosine phosphorylation following TCR stimulation. ZAP70 contains two SH2-like domains with the PTK domain located at the C-terminus. TCR-mediated Lck activity leads to phosphorylation of ZAP70 on Tyrosine 493 in the regulatory loop of the PTK domain leading to upregulation of ZAP70 kinase activity. Tyrosine 292 (Y292), in the linker region between the SH2 and PTK domains, is autophosphorylated by the activated PTK domain. By binding with c-Cbl, the phosphorylated Y292 can down-regulate TCR signaling. The J34-602 antibody recognizes ZAP70 phosphorylated at Y292.
Intracellular Staining
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Zap70 (pY292)
Monoclonal
Alexa Fluor 488
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Phosphorylated Human ZAP70
50 Tests
Cell Biology
Human
IgG1 κ
Flow Cytometry
J34-602
ZAP-70 (pY292)
Mouse
Affinity Purified
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.