missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ YPEL5 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA534351

Product Code. 15385271

  • £486.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

A suggested positive control is A-20 cell lysate. PA5-34351 can be used with blocking peptide PEP-1394.

YPEL5 (yippee-like 5) belongs to a family of five yippee-like proteins, all of which localize to the centrosome or mitotic spindle and are widely expressed in both adult and fetal tissue. This localization plus the fact that the family of human YPEL proteins share a high degree of sequence homology across species suggests that these proteins may have a conserved function involved in cell division. YPEL5 is expressed at the nucleus and centrosome during interphase; during mitosis, it localizes to the spindle poles, mitotic spindle, and spindle midzone during mitosis. Finally, during cytokinesis, YPEL5 is localized to the midbody. It is associated with the Ran Binding Protein in the Microtubule organizing center (RanBPM) and a related protein RanBP10.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

YPEL5
Polyclonal
Unconjugated
Ypel5
2310076K21Rik; CGI-127; Protein yippee-like 5; yippee like 5; yippee protein homolog; yippee-like 5; yippee-like 5 (Drosophila); Ypel5
Rabbit
Antigen affinity chromatography
RUO
383295, 51646
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Liquid
Western Blot, Immunocytochemistry
1 mg/mL
PBS with 0.02% sodium azide
P62699, P62700
Ypel5
A 14 amino acid synthetic peptide near the amino terminus of human YPEL5.
100 μg
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.