Learn More
Yes Mouse, Unlabeled, Clone: 1, BD
Mouse Monoclonal Antibody
£248.00 - £448.00
Specifications
Antigen | Yes |
---|---|
Clone | 1 |
Concentration | 250μg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15865419
|
BD Biosciences
610375 |
50 μg |
£248.00
1 set |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15875419
|
BD Biosciences
610376 |
150 μg |
£448.00
1 set |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Yes is a member of the Src family of tyrosine kinases. The cellular gene was initially identified as a homologue of v-yes, the oncogene of avian sarcoma virus Y73. At least nine different Src family members have been identified. yes, fyn, and Src are widely expressed in a variety of cell types, while the remaining members are primarily expressed in hematopoietic cells. Members of this family have several common features: they have unique N-terminal domains, they attach to cellular membranes through a myristylated N-terminus and they have homologus SH2, SH3, and catalytic domains. The common expression patterns of Yes, fyn, and Src suggest some overlapping functions. All three of these kinases can physically associate with a number of cell-surface molecules which appear to increase their catalytic activity.
Specifications
Yes | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG1 | |
Affinity Purified |
1 | |
Western Blot | |
Unconjugated | |
RUO | |
Canine, Chicken, Human, Mouse, Rat | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.