Learn More
xylulokinase homolog (H. influenzae), Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human XYLB protein.
Brand: Abnova H00009942-B01.50uL
Description
The protein encoded by this gene shares 22% sequence identity with Hemophilus influenzae xylulokinase, and even higher identity to other gene products in C. elegans (45%) and yeast (31-35%), which are thought to belong to a family of enzymes that include fucokinase, gluconokinase, glycerokinase and xylulokinase. These proteins play important roles in energy metabolism. [provided by RefSeq
Sequence: MAEHAPRRCCLGWDFSTQQVKVVAVDAELNVFYEESVHFDRDLPEFGTQGGVHVHKDGLTVTSPVLMWVQALDIILEKMKASGFDFSQVLALSGAGQQHGSIYWKAGAQQALTSLSPDLRLHQQLQDCFSISDCPVWMDSSTTAQCRQLEAAVGGAQALSCLTGSRAYERFTGNQIAKIYQQNPEAYSHTERISLVSSFAASLFLGSYSPIDYSDGSGMNLLQIQDKVWSQACLGACAPHLEEKLSPPVPSCSVVGAISSYYVQRYGFPPGCKVVAFTGDNPASLAGMRLEEGDIAVSLGTSDTLFLWLQEPMPALEGHIFCNPVDSQHYMALLCFKNGSLMREKIRNESVSRSWSDFSKALQSTEMGNGGNLGFYFDVMEITPEIIGRHRFNTENHKVAAFPGDVEVRALIEGQFMAKRIHAEGLGYRVMSKTKILATGGASHNREILQVLADVFDAPVYVIDTANSACVGSAYRAFHGLAGGTDVPFSEVVKLAPNPRLAATPSPGASQVYEALLPQYAKLEQRILSQTRGPPESpecifications
| xylulokinase homolog (H. influenzae) | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human XYLB protein. | |
| XYLB | |
| FLJ10343/FLJ12539/FLJ22075 | |
| Mouse | |
| 50 μL | |
| Kinases | |
| Primary | |
| Human | |
| Serum |
| Western Blot | |
| Unconjugated | |
| No additive | |
| NM_005108.2 | |
| XYLB | |
| XYLB (NP_005099.2, 1 a.a. ∼ 536 a.a) full-length human protein. | |
| RUO | |
| Yes | |
| 9942 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.