Learn More
XBP-1S Mouse, PE, Clone: Q3-695, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 562642
Description
The Q3-695 monoclonal antibody specifically binds to XBP-1S, the spliced isoform of X-box binding protein 1 (XBP-1). XBP-1S is generated by cells that undergo an unfolded protein response, ie, cells responding to the accumulation of unfolded proteins in the endoplasmic reticulum. It plays important roles as a transcription factor in a number of important cellular processes including the regulation of MHC class II genes, differentiation of plasma cells, immunoglobulin secretion and hepatocyte growth.
Specifications
| XBP-1S | |
| Monoclonal | |
| PE | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Human XBP-1S Recombinant Protein | |
| 50 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| Q3-695 | |
| XBP1; X-box binding protein 1; TREB5; Tax-responsive element-binding | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human, Mouse | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.