missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-XBP-1S; Clone: Q3-695; BD
Brand: BD Biosciences 562821
This item is not returnable.
View return policy
Description
Host Species: Mouse
Clone Q3-695
Isotype IgG1, κ
Species Reactivity Hu, Ms
Immunogen Human XBP-1S Recombinant Protein
Flow Cytometry, Intracellular Staining
Specifications
| XBP-1S | |
| Q3-695 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human, Mouse |
| Monoclonal | |
| Alexa Fluor 647 | |
| Human XBP-1S Recombinant Protein | |
| 50 Tests | |
| Primary | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction