Learn More
WNT3A, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human WNT3A protein.
Brand: Abnova H00089780-B01P.50ug
Description
The WNT gene family consists of structurally related genes which encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family. It encodes a protein which shows 96% amino acid identity to mouse Wnt3A protein, and 84% to human WNT3 protein, another WNT gene product. This gene is clustered with WNT14 gene, another family member, in chromosome 1q42 region. [provided by RefSeq
Sequence: MAPLGYFLLLCSLKQALGSYPIWWSLAVGPQYSSLGSQPILCASIPGLVPKQLRFCRNYVEIMPSVAEGIKIGIQECQHQFRGRRWNCTTVHDSLAIFGPVLDKATRESAFVHAIASAGVAFAVTRSCAEGTAAICGCSSRHQGSPGKGWKWGGCSEDIEFGGMVSREFADARENRPDARSAMNRHNNEAGRQAIASHMHLKCKCHGLSGSCEVKTCWWSQPDFRAIGDFLKDKYDSASEMVVEKHRESRGWVETLRPRYTYFKVPTERDLVYYEASPNFCEPNPETGSFGTRDRTCNVSSHGIDGCDLLCCGRGHNARAERRREKCRCVFHWCCYVSCQECTRVYDVHTCKNPGSRAGNSAHQPPHPQPPVRFHPPLRRAGKVPSpecifications
| WNT3A | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human WNT3A protein. | |
| WNT3A | |
| MGC119418/MGC119419/MGC119420 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 89780 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Immunofluorescence, Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC103921 | |
| WNT3A | |
| WNT3A (AAI03922, 1 a.a. ∼ 385 a.a) full-length human protein. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.