Learn More
Invitrogen™ WHSC2 Monoclonal Antibody (6B11H8)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA517199
Description
MA5-17199 targets WHSC2 in indirect ELISA, FACS, ICC, IHC, IF and WB applications and shows reactivity with Human and Rat samples. The MA5-17199 immunogen is purified recombinant fragment of human WHSC2 (amino acids: 280-511) expressed in E. Coli. MA5-17199 detects WHSC2 which has a predicted molecular weight of approximately 57.3kDa.
This gene is expressed ubiquitously with higher levels in fetal than in adult tissues. It encodes a protein sharing 93% sequence identity with the mouse protein. Wolf-Hirschhorn syndrome (WHS) is a malformation syndrome associated with a hemizygous deletion of the distal short arm of chromosome 4. This gene is mapped to the 165 kb WHS critical region, and may play a role in the phenotype of the WHS or Pitt-Rogers-Danks syndrome. The encoded protein is found to be capable of reacting with HLA-A2-restricted and tumor-specific cytotoxic T lymphocytes, suggesting a target for use in specific immunotherapy for a large number of cancer patients. This protein has also been shown to be a member of the NELF (negative elongation factor) protein complex that participates in the regulation of RNA polymerase II transcription elongation.
Specifications
| WHSC2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| Q9H3P2 | |
| NELFA | |
| Purified recombinant fragment of human WHSC2 (amino acids: 280-511) expressed in E. Coli. | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG2b |
| ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 6B11H8 | |
| Unconjugated | |
| NELFA | |
| FLJ10442; FLJ25112; LOW QUALITY PROTEIN: negative elongation factor A; mWHSC2; Negative elongation factor A; negative elongation factor complex member A; negative elongation factor complex member A, Whsc2; Nelfa; NELF-A; P/OKcl.15; Whsc2; Whsc2h; Wolf-Hirschhorn syndrome candidate 2; Wolf-Hirschhorn syndrome candidate 2 homolog; Wolf-Hirschhorn syndrome candidate 2 protein | |
| Mouse | |
| Protein G | |
| RUO | |
| 305455, 7469 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.