Learn More
Invitrogen™ WDR1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA527654
Description
Recommended positive controls: H1299, rat brain. Predicted reactivity: Mouse (97%), Rat (97%), Zebrafish (82%), Xenopus laevis (88%), Dog (99%), Chicken (94%), Chimpanzee (100%), Bovine (97%), Guinea pig (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a protein containing 9 WD repeats. WD repeats are approximately 30- to 40-amino acid domains containing several conserved residues, mostly including a trp-asp at the C-terminal end. WD domains are involved in protein-protein interactions. The encoded protein may help induce the disassembly of actin filaments. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
| WDR1 | |
| Polyclonal | |
| Unconjugated | |
| WDR1 | |
| Actin-interacting protein 1; Aip1; D5Wsu185e; epididymis secretory protein Li 52; HEL-S-52; NORI-1; rede; WD repeat domain 1; WD repeat protein 1; WD repeat-containing protein 1; WD40 repeat protein 1; Wdr1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 360950, 9948 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.89 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| O75083, Q5RKI0 | |
| WDR1 | |
| Recombinant protein encompassing a sequence within the center region of human WDR1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.