missing translation for 'onlineSavingsMsg'
Learn More

Vti1a Mouse, Unlabeled, Clone: 45, BD

Product Code. 15885839
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15885839 50 μg 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15885839 Supplier BD Biosciences Supplier No. 611220

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Eukaryotic protein trafficking involves the packaging of molecules into membranous vesicles that bud from a donor compartment, travel to a specific destination, fuse, and release their components into an acceptor compartment. Recognition between vesicle and acceptor membrane is mediated by the pairing of the integral membrane SNARE proteins. The stable interaction between vesicle proteins (v-SNAREs) and target proteins (t-SNAREs) juxtaposes the membranes and results in an activated docked state and/or membrane fusion. VTI1a and VTI1b are putative mammalian SNARE proteins identified by sequence comparison with yeast SNAREs. In line with their involvement in vesicle transport, these molecules are expressed in a wide range of mammalian tissues. Vti1a, a possible t-SNARE, contains a C-terminal hydrophobic domain and several regions that may form coiled-coil structures. It exists in distinct syntaxin 5- and syntaxin 6-containing SNARE complexes within the Golgi apparatus. Inhibition of Vti1a blocks transport of G proteins to the cell surface and results in their accumulation within the Golgi. Thus, Vti1a functions in protein transport within the secretory pathway.

Host Species: Mouse
Clone: 45
Isotype: IgG1
Species Reactivity: Rat
Immunogen: Mouse Vti1a aa.114-217
Formula Weight [Chemical]: 29kDa

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Vti1a
Applications Western Blot
Classification Monoclonal
Clone 45
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Mouse Vti1a aa.114-217
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.